missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ HiPure Expi Plasmid Gigaprep Kit

Product Code. 15565730
Change view
Click to view available options
Quantity:
2 preps
Unit Size:
250mL
Product Code. Quantity unitSize
15565730 2 preps 250mL
1 options
This item is not returnable. View return policy

Product Code. 15565730

Brand: Invitrogen™ K210009XP

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

The PureLink™ HiPure Plasmid Gigaprep Kit also includes vacuum filtration columns that provide bacterial filtration without centrifugation, enabling extremely fast process times.

The PureLink™ HiPure Plasmid Gigaprep Kit also includes vacuum filtration columns that provide bacterial filtration without centrifugation, enabling extremely fast process times. The PureLink™ HiPure Expi Plasmid Gigaprep Kit typically isolates 12 mg of high quality, ultrapure plasmid DNA with inherently low endotoxin levels from 2.5 L of bacterial culture.

Advantages of the PureLink™ HiPure Expi Plasmid Gigaprep Kit include:

  • High yield—isolate up to 14 mg of high quality plasmid DNA from a single purification using 5 L of bacterial culture volume
  • Purity—inherently low endotoxin levels (typically 0.1–1.0 EU/μg) and A260/280 >1.8, making it ideal for mammalian cell transfection
  • Simple and super-fast protocol—with vacuum assisted protocol, typical plasmid isolation in as little as 90 minutes


HiPure Expi anion exchange chromatography for plasmid purification
PureLink™ HiPure Expi plasmid purification kits use a novel, proprietary anion-exchange resin to enable the purification of the highest levels of transfection-grade plasmid DNA. The enhanced resin combines large capacity with fast flow rate, high resolution, and efficient endotoxin removal. Plasmid preparations are typically completed in less than 2 hours with purity equal to two CsCl gradients. PureLink™ HiPure Expi plasmid purification kits enable the quickest process for obtaining the highest amount of transfection-grade plasmid DNA.

For Research Use Only. Not for use in diagnostic procedures.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Equilibration Buffer
• Resuspension Buffer
• Lysis Buffer
• Precipitation Buffer
• RNase A
• Wash Buffer
• Elution Buffer
• TE Buffer
• Expi HiPure Giga Filtration Column
• Expi HiPure Giga DNA Binding Column

Store all components at room temperature upon receipt.
Format Kit
Isolation Technology Anion Exchange Resin
Sample Type Bacterial Culture
For Use With (Application) Next-Generation Sequencing, Transfection, Cloning, Sequencing, Transformation, Nucleic Acid Labeling, PCR, In Vitro Transcription
High-throughput Compatibility Manual Protocols
Prep Scale 12 mg plasmid DNA
Product Line PureLink
Product Type Plasmid GigaPrep Kit
Quantity 2 preps
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Target Plasmid DNA
Test Time 2 hr.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.