missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ Pro 96 total RNA Purification Kit

Product Code. 12055825
Change view
Click to view available options
Quantity:
2 x 1 L
384 Preps
5 x 1 L
750 mL
Product Type:
RNA Lysis Buffer
RNA Purification Kit
RNA Wash Buffer I
RNA Wash Buffer II (5X)
Unit Size:
750mL
Pack of 2
Pack of 4
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Product Type Unit Size
12055825 384 Preps RNA Purification Kit Pack of 4
11528686 750 mL RNA Lysis Buffer 750mL
10694263 2 x 1 L RNA Wash Buffer II (5X) Pack of 2
10604833 5 x 1 L RNA Wash Buffer I Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 12055825 Supplier Invitrogen™ Supplier No. 12173011A
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The PureLink Pro 96 total RNA Purification Kit provides high-throughput isolation of high-quality total RNA from the widest range of sample types and sizes.

The PureLink Pro 96 total RNA Purification Kit provides high-throughput isolation of high-quality total RNA from the widest range of sample types and sizes. Isolate high yields of total RNA with low genomic DNA contamination from bacteria, yeast, plant, or mammalian samples.

The one-hour procedure requires no handling or disposal of organic waste. RNA binds to a silica membrane and impurities are removed by washing through vacuum filtration or centrifugation. High-purity total RNA is eluted into RNase-free water; ideal for use in a variety of applications, including RT-PCR, qPCR, northern blotting, and nuclease protection assays.

Improved plate design

The PureLink Pro 96 total RNA Purification Kit provides optimized semi-skirted 96-well plates(Figure 3) for improved performance and results. In addition, the plate nozzles with annular collar prevent cross-contamination of samples and improve drying of the silica membrane to prevent ethanol carryover. In addition, sample processing can be performed manually using a benchtop vacuum manifold, by centrifugation (at 2250 x g), or with an automated liquid handling platform. The semi-skirted plate design is compatible with most vacuum manifolds on robotic workstations.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 150 mL PureLink Pro 96 Lysis Buffer; store at room temperature
• 250 mL PureLink Pro 96 Wash Buffer I; room temperature
• 250 mL PureLink Pro 96 Wash Buffer II; room temperature
• 75 mL PureLink Pro 96 RNase-free Water; room temperature
• 4 PureLink 96-well Total RNA Filter Plates; room temperature
• 4 PureLink Pro 96 Elution Plates; room temperature
Isolation Technology Silica Plate
Purification Time 25 min. (After sample homogenization)
Elution Volume 45–170 μL
Sample Type Bacteria, Cells, Plant, Tissue, Yeast
Final Product Type Total RNA
High-throughput Compatibility High-throughput Compatible
For Use With (Equipment) Biomek FX, Robotic Liquid Handling Systems, Eppendorf epMotion TECAN Freedom Evo, TECAN Genesis
For Use With (Application) RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling
Product Type RNA Purification Kit
Quantity 384 Preps
Scale Mini
Shipping Condition Room Temperature
Starting Material Amount Bacteria: ≤109 cells, Cells: varies, Plant: ≤150 mg, Tissue: ≤100 mg, Yeast: ≤1.8 mL
Yield Up to 33 μg
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • PureLink Pro 96 total RNA Purification Kit
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 1 B
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Protein denaturant,Thiocyanic acid

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.