missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ RNase A (20 mg/mL)

Product Code. 10618703
Change view
Click to view available options
Quantity:
10 mL
25 mL
Unit Size:
25mL
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10618703 10 mL Each
11508686 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10618703 Supplier Invitrogen™ Supplier No. 12091021

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

PureLink™ RNase A (ribonuclease A) is a bovine pancreatic ribonuclease that cleaves single-stranded RNA.

  • RNase A (ribonuclease A) is a bovine pancreatic ribonuclease that cleaves single-stranded RNA
  • Used for the isolation of RNA-free DNA

Concentration

  • 20 mg RNase A/mL in 50 mM Tris-HCl (pH 8.0), 10 mM EDTA

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage One 10-mL bottle

Store at 2°C to 8°C.
Concentration 20 mg/mL
Packaging Type Bottle
Product Line UltraPure
Quantity 10 mL
Shipping Condition Room Temperature
Form Liquid
I need to digest my total RNA for my ribonuclease protection assay. What RNase should I use?

Total RNA can be digested using RNAse A or RNase T1. A mix of both can be used in ribonuclease protection assays.

Is RNase A treatment necessary when isolating HMW DNA from cells or tissue samples?

The RNase A treatment is optional but highly recommended for optimal results. RNAse A treatment helps to remove RNA and single stranded DNA, increasing the purity of double stranded DNA. We recommend using the PureLink RNase A (20 mg/mL) (Cat. No. 12091021).

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.