missing translation for 'onlineSavingsMsg'
Learn More

Labcon™ PurePlus™ Cap Strips for 1.2 mL Sample Library Tubes

Product Code. 17577936
Change view
Click to view available options
Format:
Strips of 12
Strips of 8
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 125
Pack of 80
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format Sterility Unit Size
17577936 Strips of 8 Non-sterile Pack of 125
17587936 Strips of 8 Sterile Pack of 125
17507946 Strips of 12 Sterile Pack of 80
17597936 Strips of 12 Non-sterile Pack of 80
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17577936 Supplier Labcon™ Supplier No. 39138500009
  • ONLINE EXCLUSIVE PRICE
    22.85 €
    22.21 € / Pack of 125
    Save 0.64 €
    2% Off
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Complete your 1.2 mL sample library tubes with these cap strips.

  • For use with PurePlus™ 1.2 mL Sample Library Tubes

Specifications

Color Natural
Format Strips of 8
Quantity 125 Tubes/Pk.
Sterility Non-sterile
For Use With (Equipment) PurePlus™ 1.2 mL Sample Library Tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.