missing translation for 'onlineSavingsMsg'
Learn More

HALYARD™ PUREZERO™ HG3 White Nitrile Gloves
BIOPHARMA

Product Code. 17364320 Shop All Halyard Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
Unit Size:
Pack of 1000
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17364320 Large Pack of 1000
17304330 X-Small Pack of 1000
17384320 Small Pack of 1000
17374320 Medium Pack of 1000
17394320 X-Large Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 17364320 Supplier HALYARD™ Supplier No. CLN3031LG
  • ONLINE EXCLUSIVE PRICE
    305.80 €
    285.17 € / Pack of 1000
    Save 20.63 €
    6% Off
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-quality gloves designed to the exact standards to meet the requirements of a contamination controlled environment.

  • Low particulate counts and endotoxin levels due to repeated washes in deionized water and chlorination
  • Nickel-free
  • Powder-free
  • Silicone-free
  • Latex-free and no phtalates
  • High cleanliness
  • Low endotoxin levels
  • Formulated with ZERO presence of these hazardous traditional accelerators: Sulfur, Thiurams, Thiazoles, Guanidine, Carbamates(ZDEC or ZDBC)

Specifications

Certifications/Compliance ISO 3, ISO 4, ISO 5, ISO 6, ISO 7
Certifications/Compliance (Europe) GMP Grade A/B/C/D
Chemotherapeutic Rated ASTM D6978
Chlorinated Yes
Cleanroom Class Class 100 (ISO 5)
Accelerator Free Yes
AQL 1
Color White
Disposable Single-use
Exterior Finish Fully Textured
Finger Thickness (English) 6 mil
Finger Thickness (Metric) 0.16 mm
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Palm Thickness (English) 5 mil
Palm Thickness (Metric) 0.13 mm
Powdered Powder Free
Product Type Controlled Environment Gloves
Size Large
Sterility Non-sterile
Thickness (English) 6 mil
Thickness (Metric) 0.16 mm
Manufacturing/QMS Audit Standards ISO 9001:2015, EN13485:2016
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.