Description
- Anti-Scaling Softener Cartridge treats and conditions hard water
- Activated Carbon Cartridge removes particles, organics and chlorine
Specifications
Specifications
| For Use With (Equipment) | Accu500 |
| Type | Cartridge, Activated Carbon |
| Height (Metric) | 254 mm |