missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Puromycin dihydrochloride

Product Code. 18762604
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18762604 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18762604 Supplier R&D Systems™ Supplier No. 4089/50
  • ONLINE EXCLUSIVE PRICE
    157.55 €
    140.70 € / 50mg
    Save 16.85 €
    10% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Protein synthesis inhibitor

Puromycin dihydrochloride is a protein synthesis inhibitor; leads to the premature release of polypeptide chains as polypeptidyl purine derivatives. Analog of the 3' end of aminoacyl-tRNA. Aminonucleoside antibiotic. Inhibits translation in both in vitro and in vivo systems. Also inhibits the transport of proteins into the mitochondria in vitro.
TRUSTED_SUSTAINABILITY

Specifications

CAS 58-58-2
Quantity 50 mg
Product Line Tocris
Molecular Formula C22H31Cl2N7O5
Formula Weight 544.43
Product Type Puromycin Dihydrochloride
Purity >98%
Molecular Weight (g/mol) 544.43
Chemical Name or Material Puromycin dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.