missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Pursept™ A Xpress Surface Disinfecting Tissues

Product Code. p-6837048 Shop All Brand Products
Click to view available options
Container Type:
Dispenser
Refill
Unit Size:
Pack of 6
This item is not returnable. View return policy

Product Code. 10150811

Brand: BRAND™ 44840

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Simply wipe the surfaces with the tissue and leave to soak. BRAND™ Pursept™ A Xpress Tissues are effective against bacteria and fungi, with limited virucidal efficacy* against HIV, HBV and HCV. As recommended by the RKI, Bundesgesundheitsblatt 01/2004.

Simply wipe the surfaces with the tissue and leave to soak.

Ingredients:
  • 1 Pursept™ A Xpress tissue contains approx.: 1027 mg ethanol, 0.56 mg n-alkyl-aminopropyl-glycine, auxiliary agents.
Chemical and physical specifications:
  • Active solution (Pursept&;trade; A Xpress): Clear, colorless liquid, fresh smell.
  • Density: approx. 0.9 g/ml.
  • pH value: approx. 7.5.
  • Tissues: FCC-bleached (oxidation bleach), bio-degradable.
Microbiological efficiency of the undiluted working solution:
  • Surface disinfection: Bacteria and fungi with little organic burden with mechanics to DGHM/VAH: 15 seconds.
  • Limited virucidal efficacy* (incl. HIV, HBV, HCV): 15 seconds.
  • Effectiveness against TbB: 30 seconds.
* As recommended by the RKI, Bundesgesundheitsblatt 01/2004.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Disinfectant Wipe
pH 7.5
Certifications/Compliance DGHM/VAH Tested and RKI Compliant
Composition 1027mg ethanol, 0.56mg n-alkyl-aminopropyl-glycine, auxiliary agents.
Container Type Dispenser
Format Wipe
For Use With (Application) Aerosol-Free Application
Quantity 150 Tissues/Pk., 6 Pk./Cs.
compliance-icons compliance-icons
Product Identifier
  • pursept A Xpress 150 Tücher Spenderdose
Signal Word
  • Danger||Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contains : Ethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.