missing translation for 'onlineSavingsMsg'
Learn More

Push/Pull Spring Balance

Product Code. 12747056
Encompass_Preferred
Change view
Click to view available options
Capacity:
1000 g
2000 g
5000 g
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
12747056 5000 g -
12717056 1000 g -
12727056 2000 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12747056 Supplier Brand Not Defined Supplier No. 83213

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Color Yellow
For Use With (Application) For Weighing and for Measuring Force of Pushing or
Capacity 5000 g
Type Push/Pull Spring Balance
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.