missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific™ PVC Bulk Wrapped Reagent Reservoir

Product Code. 16371076 Shop All Thermo Scientific CO₂ Incubators Products
Change view
Click to view available options
Capacity (Metric):
55 mL
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
16371076 55 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16371076 Supplier Heathrow Scientific™ Supplier No. 120753
  • ONLINE EXCLUSIVE PRICE
    44.45 €
    41.96 € / Pack of 100
    Save 2.49 €
    5% Off
View all promotions
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures.

  • Trough design with a v shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
  • Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
  • Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
  • Designed to stay flat on the benchtop and not warp
  • Quality certificate available on request
  • Disposable

Specifications

Capacity (English) 1.9 oz.
Color Clear
Capacity (Metric) 55 mL
Dimensions (L x W x H) 4 x 1.10 x 1.20 in.
Material PVC
Sterility Non-sterile
Type Reagent Reservoir
For Use With (Application) Reagent Pipetting
Packaging Bulk Wrapped
Quantity 100 Pk.
Autoclavable Not Autoclavable
Channels 1
Disposable Single-use
Bottom Shape Convoluted
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.