missing translation for 'onlineSavingsMsg'
Learn More

Kautex™ PVC Captive Caps

Product Code. 13284329
Click to view available options
Thread Size (Metric):
00 mm
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 13284329

Brand: Kautex™ 2000073350

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Manufactured from PVC for durability. Kautex™ Captive Caps slide on to tapered closures with thin tips.

  • Bright red; easy to see
  • For use with closures for narrow mouth bottles
  • Available to fit closures of varying sizes
TRUSTED_SUSTAINABILITY

Specifications

Color Red
Material PVC
Thread Size (Metric) 00 mm
Neck Finish 00 mm
Product Type Cap Captive
For Use With (Equipment) Dropper Closures for Bottles with Capacity: 10/20/30/50/100/200/250/500/1000/2000/3000 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.