missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences PVC-Coated Reagent Bottles with Polypropylene Caps

Product Code. 12462571 Shop All Wheaton Science Products Products
Change view
Click to view available options
Capacity (Metric):
125 mL
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12462571 125 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12462571 Supplier DWK Life Sciences Supplier No. 221014
  • ONLINE EXCLUSIVE PRICE
    142.00 €
    129.30 € / Pack of 6
    Save 12.70 €
    8% Off
Estimated Shipment: 01-07-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Make your lab safer.

Tough coating eliminates glass scatter if broken, so cleanup is easy and safe.
  • Inert PVC coating resists strong acids, alkalis and most solvents
  • Borosilicate glass is strong and crack resistant, and no-drip pour lip offers safer, cleaner pouring
  • 4.2 and 8.45 oz. (125 and 250mL) bottles feature a special screw cap and label to help meet OSHA requirements
  • 0.6 gal. (2300mL) bottles have a finger grip

Specifications

Capacity (Metric) 125 mL
Color Clear
Diameter (English) 4.8 in.
Material Borosilicate Glass
Closure Color White
Height (Metric) 122 mm
Height (English) 4.8 in.
Closure Liner PTFE
Closure Material Polypropylene
Closure Size 33-430 mm
Diameter (Metric) 122 mm
Closure Type Screw Cap
Capacity (English) 4 oz.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.