Learn More
Saint-Gobain Performance Plastics PVDF Clamp Flow Meter
Product Code. 30284854
Change view
Click to view available options
Includes:
4.5 PVDF, 1/8 NPT Clamp
Unit Size:
Each
Product Code. Includes Unit Size
30284854 4.5 PVDF, 1/8 NPT Clamp Each
1 options
Product Code. 30284854 Supplier Saint-Gobain Performance Plastics Supplier No. 0085PHP01LA
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Enables fast wetted part exchange and accurate IR-based flow readings with gamma sterilization support
  • Infrared (IR) turbine rotor reflection results in accurate and repeatable flow measurements
  • USP Class VI PVDF material can be gamma sterilized up to 50kGy
  • Each sensor is pre-calibrated to reach <1% accuracy
  • High resolution square wave output
  • Performs an easy and fast exchange of the wetted parts
  • Small housing with screw gap to mount onto your device or system
  • Tubes can be sterilized up to 150°C (302°F)
  • Reduces down-time and prevents cross-contamination
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Flowmeter
Type Flow Sensor (Clamp-mount)
Includes 4.5 PVDF, 1/8 NPT Clamp
Airflow Rate 0.03 to 2 L/min.