missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Pyr 10
GREENER_CHOICE

Product Code. 18758536
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18758536 10 mg 10mg
18768946 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18758536 Supplier Tocris Bioscience™ Supplier No. 6941/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective TRPC3 inhibitor

Pyr 10 is a TRPC3 (transient receptor potential canonical 3) channel inhibitor (IC50 = 0.72 μM for TRPC3 mediated calcium entry in vitro), which displays approximately 18-fold selectivity for TRPC3-mediated receptor operated calcium entry (ROCE) over STIM1/Orai1-mediated store operated calcium entry (SOCE). Pyr 10 inhibits NFATc3 activation and inhibits proliferation of rat ventricular cardiac fibroblasts.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[4-[3,5-bis(Trifluoromethyl)-1 H-pyrazol-1-yl]phenyl]-4-methylbenzenesulfonamide
CAS 1315323-00-2
Quantity 10 mg
Target TRPC Channel Blockers
Molecular Formula C18H13F6N3O2S
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.