missing translation for 'onlineSavingsMsg'
Learn More

PYR-41, MedChemExpress

Product Code. 16465628
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16465628 10 mg 10mg
16456156 100 mg 100mg
16441797 50 mg 50mg
16418327 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16465628 Supplier Medchem Express Supplier No. HY13296
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress PYR-41 is a selective and cell permeable inhibitor of ubiquitin-activating enzyme E1 with an IC50 of < 10 μM, with little activity at E2 and E3.

PYR-41 is a selective and cell permeable inhibitor of ubiquitin-activating enzyme E1 with an IC50 of < 10 μM, with little activity at E2 and E3.

Specifications

Chemical Name or Material PYR-41
CAS 418805-02-4
Color Red browm
Purity Grade Notes Research
Molecular Formula C17H13N3O7
Quantity 10 mg
Solubility Information DMSO : ≥ 46 mg/mL (123.89 mM)
SMILES O=C(N1)/C(C(N1C2=CC=C(C=C2)C(OCC)=O)=O)=C/C3=CC=C([N+]([O-])=O)O3
Molecular Weight (g/mol) 371.3
Formula Weight 371.3
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.