missing translation for 'onlineSavingsMsg'
Learn More

PYR-41, TRC

Product Code. 30409535
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30409535 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30409535 Supplier Toronto Research Chemicals Supplier No. TRCP8403805MG
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material ethyl 4-[(4Z)-4-[(5-nitro-2-furyl)methylene]-3,5-dioxo-pyrazolidin-1-yl]benzoate
CAS 418805-02-4
Molecular Formula C17 H13 N3 O7
Quantity 5 mg
Synonym 4-[4-[(5-Nitro-2-furanyl)methylene]-3,5-dioxo-1-pyrazolidinyl]-benzoic acid Ethyl Ester, PYR-41
InChI Formula InChI=1S/C17H13N3O7/c1-2-26-17(23)10-3-5-11(6-4-10)19-16(22)13(15(21)18-19)9-12-7-8-14(27-12)20(24)25/h3-9H,2H2,1H3,(H,18,21)/b13-9-
SMILES CCOC(=O)c1ccc(cc1)N2NC(=O)\C(=C\c3oc(cc3)[N+](=O)[O-])\C2=O
IUPAC Name ethyl 4-[(4Z)-4-[(5-nitro-2-furyl)methylene]-3,5-dioxo-pyrazolidin-1-yl]benzoate
Molecular Weight (g/mol) 371.3
Formula Weight 371.0753
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.