missing translation for 'onlineSavingsMsg'
Learn More

Pyrazine-2-carboxylic acid, 99%

Product Code. 11480993
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11480993 100 g 100g
11470993 25 g 25g
11490993 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480993 Supplier Thermo Scientific Alfa Aesar Supplier No. A13363.22
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 98-97-5 | C5H4N2O2 | 124.099 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-97-5
Molecular Formula C5H4N2O2
Molecular Weight (g/mol) 124.099
MDL Number MFCD00006130
InChI Key NIPZZXUFJPQHNH-UHFFFAOYSA-N
Synonym 2-pyrazinecarboxylic acid, pyrazinoic acid, pyrazinecarboxylic acid, pyrazinic acid, 2-carboxypyrazine, paradiazinecarboxylic acid, pyrazinemonocarboxylic acid, 2-pyrazinoic acid, 2-pyrazine carboxylic acid, piazinecarboxylic acid
PubChem CID 1047
ChEBI CHEBI:71311
IUPAC Name pyrazine-2-carboxylic acid
SMILES C1=CN=C(C=N1)C(=O)O

Specifications

Melting Point ∼225°C (decomposition)
Quantity 100 g
Beilstein 112305
Merck Index 14,7959
Formula Weight 124.1
Percent Purity 99%
Chemical Name or Material Pyrazine-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.