missing translation for 'onlineSavingsMsg'
Learn More

Pyrazine, 99+% (dry wt.) water <1.0%

Product Code. 11481738
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11481738

Brand: Thermo Scientific Alfa Aesar A13982.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 290-37-9 | C4H4N2 | 80.09 g/mol

Pyrazine is used in flavoring, fragrance, and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrazine is used in flavoring, fragrance, and dyes.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 290-37-9
Molecular Formula C4H4N2
Molecular Weight (g/mol) 80.09
MDL Number MFCD00006122
InChI Key KYQCOXFCLRTKLS-UHFFFAOYSA-N
Synonym 1,4-diazine, p-diazine, paradiazine, piazine, 1,4-diazabenzene, pyrazin, ccris 1331, 1,4-diazin, mixture, mu-pyrazine;
PubChem CID 9261
ChEBI CHEBI:30953
IUPAC Name pyrazine
SMILES C1=CN=CC=N1

Specifications

Melting Point 50°C to 56°C
Boiling Point 113°C to 116°C
Flash Point 55°C (131°F)
Odor Strong
Quantity 5 g
UN Number UN1325
Beilstein 103905
Merck Index 14,7957
Formula Weight 80.09
Percent Purity ≥99%
Assay (dry wt.) water <1.0%
Chemical Name or Material Pyrazine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.