missing translation for 'onlineSavingsMsg'
Learn More

Pyrazine, 99+%

Product Code. 11966601 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11966601 100 g Glass bottle 100g
10377160 10 g Glass bottle 10g
10735621 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11966601 Supplier Thermo Scientific Acros Supplier No. 131701000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 290-37-9 | C4H4N2 | 80.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 290-37-9
Molecular Formula C4H4N2
Molecular Weight (g/mol) 80.09
InChI Key KYQCOXFCLRTKLS-UHFFFAOYSA-N
Synonym 1,4-diazine, p-diazine, paradiazine, piazine, 1,4-diazabenzene, pyrazin, ccris 1331, 1,4-diazin, mixture, mu-pyrazine;
PubChem CID 9261
ChEBI CHEBI:30953
IUPAC Name pyrazine
SMILES C1=CN=CC=N1

Specifications

Melting Point 52.0°C to 56.0°C
Color White
Boiling Point 115.0°C to 116.0°C
Flash Point 55°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 100 g
Beilstein 23,91
Merck Index 15,8068
Solubility Information Solubility in water: freely soluble. Other solubilities: freely soluble in alcohol and ether
Formula Weight 80.09
Percent Purity 99+%
Physical Form Crystals
Chemical Name or Material Pyrazine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Pyrazine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.