missing translation for 'onlineSavingsMsg'
Learn More

Pyrazole, 98%, pure

Product Code. 10235280 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10235280 100 g Glass bottle 100g
10225280 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10235280 Supplier Thermo Scientific Acros Supplier No. 131741000
 Restricted Item
In Stock
Læg i indkøbskurv
Læg i indkøbskurv
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

CAS: 288-13-1 | C3H4N2 | 68.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 288-13-1
Molecular Formula C3H4N2
Molecular Weight (g/mol) 68.08
MDL Number MFCD00005234
InChI Key WTKZEGDFNFYCGP-UHFFFAOYSA-N
Synonym pyrazole, 1,2-diazole, 1h-pyrazol, pyrazol, unii-3qd5kjz7zj, 3qd5kjz7zj, diazole, hpz, 1-h-pyrazole, pyrazol#1
PubChem CID 1048
ChEBI CHEBI:17241
IUPAC Name 1H-pyrazole
SMILES C1=CNN=C1

Tekniske data

Melting Point 66.0°C to 70.0°C
Color White to Yellow
Boiling Point 186.0°C to 188.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 100 g
Beilstein 23, 39
Fieser 01,958; 12,419
Merck Index 15, 8071
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,ether and benzene
Formula Weight 68.08
Percent Purity 98%
Grade Pure
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Pyrazole, 98%
Vis mere Vis mindre
compliance-icons
Produktidentifikator
  • Pyrazole
Signalord
  • Warning
Farekategori
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faresætning
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Sikkerhedssætning
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.