missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ PYREX™ Disposable Bulk Glass Serological Pipets, TD, Non-Sterile, Unplugged
Shop All Corning ProductsDescription
These disposable glass pipets are calibrated 'To Deliver' with blow-out and offer long, slender, tapered tips to make pipetting go faster and easier. All pipets have negative graduations and fire-polished tips for burr-free, even flow rates. Nonsterile, unplugged, and bulk packed. Reference: ASTM E-714.
Tekniske data
Tekniske data
| Aspirator Plug | No |
| Calibrated | Yes |
| Calibration | To Deliver |
| Certifications/Compliance | ASTM E714 |
| Material | Glass |
| Sterility | Non-sterile |
| For Use With (Application) | Diagnostic, Blood, Serum, |
| Graduated | Yes |
| Graduations | 0.1 mL |
| Graduation Type | Negative Graduations |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?