missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Borosilicate Glass Erlenmeyer Flask with Quickfit Ground Glass Socket

Product Code. 12622062
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
Standard Taper Joint:
19/26
24/29
29/32
Unit Size:
Pack of 10
Pack of 20
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Standard Taper Joint unitSize
12622062 250 mL 29/32 Pack of 10
12463431 500 mL 29/32 Pack of 10
12453431 250 mL 24/29 Pack of 10
15597660 50 mL 19/26 Pack of 20
15507670 100 mL 29/32 Pack of 20
15517670 1000 mL 29/32 Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 12622062 Supplier Pyrex Supplier No. 1132/15
Add to basket
Add to basket
This item is not returnable. View return policy

Grow cultures and store other laboratory reagents in Glass Erlenmeyer flasks. The Pyrex™ borosilicate glass Erlenmeyer flask with QuickFit™ glass socket is sturdy and comes in a variety of sizes.

Grow cultures and store other laboratory reagents in Glass Erlenmeyer flasks. The Pyrex™ borosilicate glass Erlenmeyer flask with QuickFit™ glass socket is sturdy and comes in a variety of sizes.

  • Complies to ISO 4797
  • Conical flasks with highest quality Quickfit™ ground glass socket
  • Borosilicate glass for corrosion free performance and high resistance to thermal shock
  • Erlenmeyer flasks with high quality Quickfit™ ground glass socket
  • Manufactured from Pyrex™ borosilicate glass for highest resistance to thermal shock and chemical corrosion
  • White enamel graduations and marking spot
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance DIN 12387
Capacity (Metric) 250 mL
Material Borosilicate Glass
Graduations Yes
Height (Metric) 130 mm
Shape Conical
Standard Taper Joint 29/32
Writing Surface Yes
Graduated Yes
Diameter (Metric) Outer 85 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.