Learn More
Pyrex™ Borosilicate Glass Graduated Test Tubes with Stopper
Manufactured with borosilicate glass for excellent chemical and thermal shock resistance. Pyrex™ Borosilicate Glass Graduated Test Tubes with Stopper are ideal for measuring and mixing liquids and solutions.
Brand:  Pyrex 1628/10M
Product Code. 11925909
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • White enamel graduations for visual volume assessment
  • Polyethylene stopper prevents spills and leakage