missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Borosilicate Glass Narrow Mouth Reagent Bottles with Glass Stopper

Product Code. 11307864
Change view
Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
20,000 mL
2000 mL
250 mL
500 mL
5000 mL
Unit Size:
Each
Pack of 10
Pack of 30
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11307864 500 mL Pack of 10
12643292 100 mL Pack of 10
12653292 250 mL Pack of 10
11397854 250 mL Each
11317864 1000 mL Pack of 10
15537690 2000 mL Pack of 30
11337864 5000 mL Each
11347864 10,000 mL Each
11357864 20,000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 11307864 Supplier Pyrex Supplier No. 1521/08D
Trusted Glassware Deals
Use promo code 28509
View all promotions
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Store reagents, analytical standards and culture media in these glass reagent bottles. Pyrex™ Borosilicate Glass Narrow Mouth Reagent Bottles with Glass Stopper features excellent chemical resistance and can withstand high temperatures.

  • Complies with ISO 4796-2
  • Supplied with solid glass stopper
  • Pyrex™ borosilicate glass with ground neck
  • Autoclavable
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 500 mL
Diameter (Metric) Inner Neck 24 mm
Material Borosilicate Glass
No. per Pack 10/PK
Closure Included Yes
Mouth Narrow
Height (Metric) 175 mm
Closure Material Glass
Closure Size 24/29 mm
Diameter (Metric) 89 mm
Closure Type Stopper
Certifications/Compliance ISO 4796-2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.