missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Borosilicate Glass Narrow Mouth Reagent Bottles with Glass Stopper
Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
20,000 mL
2000 mL
250 mL
500 mL
5000 mL
missing translation for 'unitSize'
Each
Pack of 10
Pack of 30
Description
- Complies with ISO 4796-2
- Supplied with solid glass stopper
- Pyrex™ borosilicate glass with ground neck
- Autoclavable
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Diameter (Metric) Inner Neck | 19 mm |
| Material | Borosilicate Glass |
| No. per Pack | 10/PK |
| Certifications/Compliance | ISO 4796-2 |
| Closure Included | Yes |
| Mouth | Narrow |
| Height (Metric) | 143 mm |
| Closure Material | Glass |
| Closure Size | 19/26 mm |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction