missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Borosilicate Glass Reagent Bottles
Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
15,000 mL
20,000 mL
2000 mL
25 mL
250 mL
50 mL
500 mL
5000 mL
Unit Size:
Each
Pack of 10
Pack of 5
Description
Store reagents, analytical standards and culture media in these borosilicate glass reagent bottles. PyrexTM Borosilicate Glass Reagent Bottles withstand hot air sterilization up to 180°C, and have excellent chemical resistance.
- Complies with ISO 4796-1
- Manufactured from Pyrex™ borosilicate glass
- Excellent resistance to chemical attack and high temperatures
- Supplied with PBT screw caps that have a PTFE faced sealing disc
- Autoclavable
- White graduations and marking spot
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| For Use With (Application) | Ideal for storage of reagents, analytical standards and culture media etc |
| Material | Borosilicate Glass |
| No. per Pack | 10/PK |
| Closure Color | Black |
| Certifications/Compliance | ISO 4796-1 |
| Closure Included | Yes |
| Graduated | Yes |
| Height (Metric) | 135 mm |
| Closure Liner | PTFE |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction