missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Borosilicate Glass Reagent Bottles with Polypropylene Cap and Pouring Ring
Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
15,000 mL
150 mL
20,000 mL
2000 mL
25 mL
250 mL
3500 mL
50 mL
500 mL
5000 mL
Unit Size:
Each
Pack of 10
Description
Mix and store a wide range of culture mediums with these glass reagent bottles. PyrexTM Borosilicate Glass Reagent Bottles with Polypropylene Cap and Pouring Ring feature a high chemical resistance and are designed with sloped shoulders for smooth pouring.
- Bottles complete with polypropylene cap and drip-free pouring ring
- Complies with ISO 4796
- Printed with Trace Code for downloadable batch certificate
- Sloping shoulders for smooth pouring action
- White graduations and marking spot
- Manufactured from temperature and chemically resistant borosilicate glass
- For use up to 140°C
Spécification
Spécification
| Capacity (Metric) | 2000 mL |
| Capacity (English) | 0.5 gal. |
| Color | Clear |
| Diameter (English) | 5.4 in. |
| For Use With (Application) | Ideal for mixing and storage of culture medium |
| Includes | Polypropylene screw cap and pouring ring |
| Material | Borosilicate Glass |
| No. per Pack | 10/PK |
| Quantity | 10 Pk. |
| Thread Size (Metric) | GL 45 |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu