Learn More
PYREX™ Culture Flask Closure
Product Code. 12687266
Change view
Click to view available options
Closure Material:
Aluminum
Polypropylene
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Material Unit Size
12687266 Aluminum Pack of 10
12524917 Polypropylene Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12687266 Supplier Pyrex Supplier No. 4550/38M
  • ONLINE EXCLUSIVE PRICE
    155.00 €
    142.08 € / Pack of 10
    Save 12.92 €
    8% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use with 1135 culture flasks, 1138/10 Fernbach flasks and 1141 baffled flasks.
  • Two types of closure - Anodised aluminum - Two-position polypropylene
  • Suitable for autoclaving
Specifications
Closure Material Aluminum
No. per Pack 10
Autoclavable Autoclavable