Learn More
Pyrex™ Gas Distribution Tube
Disperse gas in liquids to produce the optimum bubble size.  PyrexTM gas distribution tubes come in a variety of porosities.
Brand:  Pyrex 3830/04M
Product Code. 11338024
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Ideal for the dispersion of gas in liquids
  • Borosilicate glass tube with sintered thimble
  • Choice of thimble porosities to achieve optimum gas bubble size