missing translation for 'onlineSavingsMsg'
Learn More

Corning™ PYREX™ GL45 Screw Cap with Plug Seal

Product Code. 17223484 Shop All Corning Products
Change view
Click to view available options
Closure Material:
PBT
Polypropylene
Closure Color:
Orange
Red
Unit Size:
Pack of 10
Pack of 20
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Material Closure Color Unit Size
17223484 PBT Red Pack of 10
11941831 Polypropylene Orange Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17223484 Supplier Corning™ Supplier No. 139545HTC
  • ONLINE EXCLUSIVE PRICE
    118.00 €
    109.75 € / Pack of 10
    Save 8.25 €
    6% Off
Estimated Shipment: 02-04-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Nonsterile, reusable caps

These are various sizes and types of screw caps for use with storage bottles.

TRUSTED_SUSTAINABILITY

Specifications

Closure Color Red
Closure Material PBT
Color Red
For Use With (Equipment) Graduated Erlenmeyer flasks and media bottles
No. per Case 10/Cs.
No. per Pack 10/Pk.
Product Type Cap
Temperature Range (English) Up to 356°F
Temperature Range (Metric) Up to 180°C
Thread Size GL 45
Autoclavable Autoclavable
Disposable Reusable
Sterility Non-sterile
Closure Type High temperature
Show More Show Less
How can I increase the lifespan of my PYREX and PYREXPLUS bottles?

Proper care includes:
• Avoiding mechanical damage (e.g., scratches or chips)
• Preventing thermal shock
• Following recommended cleaning and sterilization techniques

How should I clean PYREX bottles?

1. Wash bottles promptly after use, or soak them in water if immediate cleaning isn’t possible
2. Sterilize bottles contaminated with biological materials before cleaning
3. Use detergent and rinse thoroughly with tap water, followed by deionized or distilled water
4. For new bottles used in cell culture, soak in a 1% hydrochloric or nitric acid solution before washing

What precautions should I take when autoclaving PYREX bottles?

• Do not tighten caps immediately after autoclaving to prevent vacuum breakage, except for vented caps (e.g., 1395-45LTMC)
• Replace vented caps after 3-5 autoclave cycles or if the membrane is damaged

Can I use PYREX bottles with bottle-top filter units?

Do not use bottle-top filter units with reagent bottles larger than 1000 mL or laboratory bottles larger than 2000 mL.

Are PYREX bottles suitable for vacuum or pressure applications?

PYREX bottles larger than 2000 mL are not recommended for vacuum applications. Additionally, 1396 and 1397 series bottles are not designed for vacuum use.

What safety precautions should I take when using PYREX bottles?

Follow these safety guidelines:
• Avoid using scratched, cracked, or nicked glassware
• Do not expose bottles to direct heat, such as a flame or hot plate
• Prevent thermal shock by avoiding rapid temperature changes (e.g., placing hot bottles on cold surfaces)
• Do not refrigerate below -20°C or expose PYREXPLUS bottles to temperatures above 80°C

Are vented caps available, and what are they used for?

Yes, vented caps with a 0.22 µm PTFE membrane allow automatic gas exchange during autoclaving. They are suitable for use with bottle sizes ranging from 100 mL to 2000 mL.

Can the caps withstand high temperatures?

The standard orange polypropylene caps can withstand up to 140°C. Optional red PBT caps with PTFE-coated liners are available for applications requiring temperatures up to 180°C.

What types of caps are available for PYREX bottles?

A variety of optional closures are available, including:
• Plug seal, GL45 and GLS80 color-coded caps for sorting and identification
• High-temperature PBT caps with PTFE-coated liners, ETFE copolymer rings, or silicone septa (suitable for dry heat sterilization up to 180°C)
• Vented GL45 caps with 0.22 µm PTFE hydrophobic membranes for automatic gas exchange
• Open-top PBT caps in GL25, GL32, and GL45 sizes
• Silicone or PTFE-faced silicone septa for open-top caps

What material are PYREX bottles made of?

PYREX bottles are made from Type I, Class A borosilicate glass, which is chemically resistant and has low thermal expansion. They come with clear polypropylene pouring rings and caps and are fully autoclavable.

What are PYREX and PYREXPLUS reusable laboratory bottles used for?

PYREX bottles are heavy-duty storage vessels designed for reagents, sterile tissue culture media, biological fluids, aqueous and nonaqueous solutions. PYREXPLUS bottles have a PVC coating for added protection, and Low Actinic bottles are available for light-sensitive materials.


Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.