missing translation for 'onlineSavingsMsg'
Learn More

Corning™ PYREX™ Media Bottle High-Temperature Caps, Open-Top

Product Code. 17263474 Shop All Corning Products
Change view
Click to view available options
Thread Size (Metric):
GL 25
GL 32
GL 45
Unit Size:
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size (Metric) Unit Size
17263474 GL 32 Pack of 10
17233474 GL 25 Pack of 10
11759455 GL 45 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17263474 Supplier Corning™ Supplier No. 139532HTSC
  • ONLINE EXCLUSIVE PRICE
    67.60 €
    63.15 € / Pack of 10
    Save 4.45 €
    6% Off
Estimated Shipment: 18-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

With drip-free pouring ring

These are various sizes and types of screw caps for use with storage bottles.

TRUSTED_SUSTAINABILITY

Specifications

Color Red
Height (Metric) With Closure 24.2 mm
Material PBT
No. per Case 10/Cs.
No. per Pack 10/Pk.
Sterility Non-sterile
Thread Size (Metric) GL 32
Closure Color Red
For Use With (Equipment) Storage bottle
Closure Material PBT
Closure Type High Temperature
Product Type Open Top Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.