missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Plain Arms, Straight Bore Stopcock

Product Code. 11372115
Change view
Click to view available options
Diameter (Metric) Side Arm:
8 mm
9 mm
12.5 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Side Arm Unit Size
11372115 9 mm Each
11362115 8 mm Each
11352115 12.5 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11372115 Supplier Pyrex Supplier No. SSD104
  • ONLINE EXCLUSIVE PRICE
    66.75 €
    61.88 € / Pack of 1
    Save 4.87 €
    7% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Externally operated valve to regulate the flow of a liquid.

  • Glass stopcocks which meet specifications of DIN 12541
  • Manufactured from borosilicate glass with interchangeable key
  • Leakage rate meets the requirements of DIN 12540 i.e. 0.1mbar l/s-1
  • Supplied complete with key retaining device

Specifications

Type Straight Bore Stopcock
Material Borosilicate Glass
Diameter (Metric) Side Arm 9 mm
Certifications/Compliance DIN 12541
Includes Key retaining device
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.