Learn More
Pyrex™ Roux Culture Off-Set Neck Flasks
Ideal for cell culture
423.00 € - 615.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
12574257
|
Pyrex
1350/06M |
1200 mL |
615.00 €
Pack of 10 |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
13411859
|
Pyrex
1350/04M |
600 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Manufactured from chemically inert Pyrex glass for a contamination-free environment
- Withstands repeated autoclaving