missing translation for 'onlineSavingsMsg'
Learn More

pyridazin-4-ylmethanol, TRC

Product Code. 30412441
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30412441 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30412441 Supplier Toronto Research Chemicals Supplier No. TRCP993368100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material pyridazin-4-ylmethanol
CAS 50901-43-4
Molecular Formula C5 H6 N2 O
Quantity 100 mg
Synonym 4-Hydroxymethylpyridazine
InChI Formula InChI=1S/C5H6N2O/c8-4-5-1-2-6-7-3-5/h1-3,8H,4H2
SMILES OCc1ccnnc1
IUPAC Name pyridazin-4-ylmethanol
Molecular Weight (g/mol) 110.114
Formula Weight 110.048
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.