missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-2,6-dicarbonyl dichloride, 97%

Product Code. 11496010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496010 25 g 25g
11476010 1 g 1g
11486010 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11496010 Supplier Thermo Scientific Alfa Aesar Supplier No. H26570.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3739-94-4 | C7H3Cl2NO2 | 204.006 g/mol

2,6-Pyridinedicarbonyl dichloride is used as the reagent during the synthesis of pyridine-based polyamido-polyester optically active macrocycles. It is used as starting reagent during the synthesis of pyridine-bridged 2,6-bis-carboxamide Schiff?s bases. It was used in the synthesis of N,N?-bis(1-pyrenylmethyl)pyridine-2,6-dicarboxamide3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Pyridinedicarbonyl dichloride is used as the reagent during the synthesis of pyridine-based polyamido-polyester optically active macrocycles. It is used as starting reagent during the synthesis of pyridine-bridged 2,6-bis-carboxamide Schiff′s bases. It was used in the synthesis of N,N′-bis(1-pyrenylmethyl)pyridine-2,6-dicarboxamide3

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3739-94-4
Molecular Formula C7H3Cl2NO2
Molecular Weight (g/mol) 204.006
MDL Number MFCD00006289
InChI Key GWHOGODUVLQCEB-UHFFFAOYSA-N
Synonym 2,6-pyridinedicarbonyl dichloride, 2,6-pyridinedicarboxylic acid chloride, pyridine-2,6-dicarbonyl dichloride, 2,6-pyridinedicarbonyl chloride, pyridine-2,6-dicarboxylic acid chloride, 2,6-pyridinedicarboxylicacidchloride, pyridine-2,6-dicarboyl dichloride, 2,6-pyridinedicarbony chloride, pubchem13243, tpc-i153
PubChem CID 77335
IUPAC Name pyridine-2,6-dicarbonyl chloride
SMILES C1=CC(=NC(=C1)C(=O)Cl)C(=O)Cl

Specifications

Melting Point 57°C to 61°C
Boiling Point 284°C
Odor Irritating
Quantity 25 g
UN Number UN3261
Beilstein 131556
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 204.01
Percent Purity 98%
Chemical Name or Material Pyridine-2,6-dicarbonyl dichloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.