missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-2-carboxaldehyde 4-nitrophenylhydrazone, 98%, Thermo Scientific™

Product Code. 11445817
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11445817 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11445817 Supplier Thermo Scientific Alfa Aesar Supplier No. B24380.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70421-66-8
Molecular Formula C12H10N4O2
Molecular Weight (g/mol) 242.24
MDL Number MFCD00012100
InChI Key JXFHYUCJPGWCFI-NTEUORMPSA-N
Synonym pyridine-2-carboxaldehyde 4-nitrophenylhydrazone, 2-pyridinecarboxaldehyde 4-nitrophenylhydrazone, pyridine-2-carbaldehyde 4'-nitrophenylhydrazone, 2-4-nitrobenzenecarbohydrazonoyl pyridine, picolinaldehyde 4-nitrophenylhydrazone, pyridine-2-carbaldehyde 4-nitrophenylhydrazone, 4-nitro-n-e-2-pyridinylmethylideneamino aniline, 4-nitro-n-e-pyridin-2-ylmethylideneamino aniline, 1e-2-2-pyridyl-1-azavinyl 4-nitrophenyl amine
PubChem CID 9562617
IUPAC Name 2-[(E)-[2-(4-nitrophenyl)hydrazin-1-ylidene]methyl]pyridine
SMILES [O-][N+](=O)C1=CC=C(N\N=C\C2=CC=CC=N2)C=C1

Specifications

Melting Point 256°C to 259°C
Quantity 25 g
Formula Weight 242.24
Percent Purity 98%
Chemical Name or Material Pyridine-2-carboxaldehyde 4-nitrophenylhydrazone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.