missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-2-carboximidamide hydrochloride, 97%, Thermo Scientific™

Product Code. 10614395
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10614395 25 g Amber Glass Bottle 25g
10069251 1 g Amber Glass Bottle 1g
10242452 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10614395 Supplier Thermo Scientific Maybridge Supplier No. RF01042EB
 Restricted Item
Estimated Shipment: 11-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 51285-26-8
Molecular Formula C6H8ClN3
Molecular Weight (g/mol) 157.601
MDL Number MFCD00052271
InChI Key GMHCEDDZKAYPLB-UHFFFAOYSA-N
Synonym picolinimidamide hydrochloride, pyridine-2-carboximidamide hydrochloride, 2-amidinopyridine hydrochloride, pyridine-2-carboxamidine hydrochloride, picolinamidine hydrochloride, picolinamidine hcl, picolinimidamide hcl, pyridine-2-carboximidamide, hcl, 2-amidinopyridine, 2-amidinopyridinium chloride
PubChem CID 12403615
IUPAC Name pyridine-2-carboximidamide;hydrochloride
SMILES C1=CC=NC(=C1)C(=N)N.Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 25 g
Formula Weight 157.6
Percent Purity 97%
Chemical Name or Material Pyridine-2-carboximidamide hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.