missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-2-thiocarboxamide, 97+%

Product Code. 15488157
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15488157 5 g 5g
15498157 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15488157 Supplier Thermo Scientific Alfa Aesar Supplier No. H51710.06
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 5346-38-3 | C6H6N2S | 138.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5346-38-3
Molecular Formula C6H6N2S
Molecular Weight (g/mol) 138.19
MDL Number MFCD00087576
InChI Key HYKQYVSNFPWGKQ-UHFFFAOYSA-N
Synonym thiopicolinamide, 2-pyridinecarbothioamide, pyridine-2-thioamide, pyridine-2-carbothioic acid amide, picolinamide, thio, 2-pyridinethiocarboxamide, picolinothioamide, 2-thiopicolinamide, pyridinecarbothioamide, 2-thiocarbamoylpyridine
PubChem CID 1549499
IUPAC Name pyridine-2-carbothioamide
SMILES NC(=S)C1=CC=CC=N1

Specifications

Melting Point 120°C to 125°C
Quantity 5 g
Formula Weight 138.19
Percent Purity ≥97%
Chemical Name or Material Pyridine-2-thiocarboxamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.