missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-3,4-dicarboxylic acid, 98%

Product Code. 11491978
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11491978 10 g 10g
11401988 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11491978 Supplier Thermo Scientific Alfa Aesar Supplier No. A14580.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 490-11-9 | C7H5NO4 | 167.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 490-11-9
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00006392
InChI Key MUYSADWCWFFZKR-UHFFFAOYSA-N
Synonym 3,4-pyridinedicarboxylic acid, cinchomeronic acid, chinchomeronic acid, 3,4-pyridinedicarboxylicacid, pyridine-3,4-dicarboxylicacid, unii-7389uzw5dy, 3,4-pyridindicarbons, cinchomeronsaure, pubchem2419, 3,4-dicarboxypyridine
PubChem CID 10273
ChEBI CHEBI:46860
IUPAC Name pyridine-3,4-dicarboxylic acid
SMILES OC(=O)C1=CC=NC=C1C(O)=O

Specifications

Melting Point ∼255°C (decomposition)
Quantity 10 g
Beilstein 137242
Merck Index 14,2283
Formula Weight 167.12
Percent Purity 98%
Chemical Name or Material Pyridine-3,4-dicarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.