missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-3-sulfonyl chloride hydrochloride, 95%, Thermo Scientific Chemicals

Product Code. 15790879 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15790879 5 g 5g
15700889 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15790879 Supplier Thermo Scientific Acros Supplier No. 451480050
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42899-76-3
Molecular Formula C5H4ClNO2S·ClH
Molecular Weight (g/mol) 214.07
InChI Key VIVPWOMJFLICOZ-UHFFFAOYSA-N
Synonym pyridine-3-sulfonyl chloride hydrochloride, pyridine-3-sulfonyl chloride hcl, 3-pyridinesulfonyl chloride hydrochloride, pyridine-3-sulphonyl chloride hydrochloride, 3-pyridinesulfonyl chloride, hcl, 3-pyridinesulfonyl chloride, hydrochloride, 3-pyridylsulfonyl chloride hydrochloride, pyridine-3-sulfonylchloride hydrochloride, pyridin-3-ylsulfonyl chloride hydrochloride, 3-pyridylsulfonylchloride hcl
PubChem CID 12571517
IUPAC Name pyridine-3-sulfonyl chloride;hydrochloride
SMILES C1=CC(=CN=C1)S(=O)(=O)Cl.Cl

Specifications

Melting Point 144.0°C to 145.0°C
Elemental Analysis 26.37 to 29.74% (C) 2.21 to 2.50% (H) 6.15 to 6.94% (N)
Infrared Spectrum Authentic
Assay Percent Range 94% min. as diethylsulfonamide (HPLC)
Quantity 5 g
Formula Weight 214.07
Percent Purity 95%
Chemical Name or Material Pyridine-3-sulfonyl chloride hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.