missing translation for 'onlineSavingsMsg'
Learn More

pyridine-3-sulfonyl chloride hydrochloride, 95%, Thermo Scientific™

Product Code. 10634385
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10634385 10 g 10g
10620895 250 mg 250mg
10104542 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10634385 Supplier Thermo Scientific Maybridge Supplier No. CC04103EA
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 42899-76-3
Molecular Formula C5H5Cl2NO2S
Molecular Weight (g/mol) 214.06
InChI Key VIVPWOMJFLICOZ-UHFFFAOYSA-N
Synonym pyridine-3-sulfonyl chloride hydrochloride, pyridine-3-sulfonyl chloride hcl, 3-pyridinesulfonyl chloride hydrochloride, pyridine-3-sulphonyl chloride hydrochloride, 3-pyridinesulfonyl chloride, hcl, 3-pyridinesulfonyl chloride, hydrochloride, 3-pyridylsulfonyl chloride hydrochloride, pyridine-3-sulfonylchloride hydrochloride, pyridin-3-ylsulfonyl chloride hydrochloride, 3-pyridylsulfonylchloride hcl
PubChem CID 12571517
IUPAC Name pyridine-3-sulfonyl chloride;hydrochloride
SMILES C1=CC(=CN=C1)S(=O)(=O)Cl.Cl

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 214.07
Percent Purity 95%
Chemical Name or Material Pyridine-3-sulfonyl chloride hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.