missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Pyridine-3-sulfonyl chloride hydrochloride, 97%

Product Code. 15450898
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15450898 5 g 5g
15440898 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15450898

Brand: Alfa Aesar™ H54672.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Reacts with water.

Notes
Store at -20°C. Keep away from light and heat. Hygroscopic. Store under nitrogen.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42899-76-3
Molecular Formula C5H5Cl2NO2S
Molecular Weight (g/mol) 214.06
MDL Number MFCD04035552
InChI Key VIVPWOMJFLICOZ-UHFFFAOYSA-N
Synonym pyridine-3-sulfonyl chloride hydrochloride, pyridine-3-sulfonyl chloride hcl, 3-pyridinesulfonyl chloride hydrochloride, pyridine-3-sulphonyl chloride hydrochloride, 3-pyridinesulfonyl chloride, hcl, 3-pyridinesulfonyl chloride, hydrochloride, 3-pyridylsulfonyl chloride hydrochloride, pyridine-3-sulfonylchloride hydrochloride, pyridin-3-ylsulfonyl chloride hydrochloride, 3-pyridylsulfonylchloride hcl
PubChem CID 12571517
IUPAC Name pyridine-3-sulfonyl chloride;hydrochloride
SMILES C1=CC(=CN=C1)S(=O)(=O)Cl.Cl

Specifications

Quantity 5 g
UN Number UN3261
Beilstein 3706102
Formula Weight 176.97
Percent Purity 97%
Chemical Name or Material Pyridine-3-sulfonyl chloride hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.