missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-4-boronic acid, 97%, Thermo Scientific™

Product Code. 10503153
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10503153 10 g Amber Glass Bottle 10g
10337853 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10503153 Supplier Thermo Scientific Maybridge Supplier No. CC04212EA
 Restricted Item
Estimated Shipment: 13-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1692-15-5
Molecular Formula C5H6BNO2
Molecular Weight (g/mol) 122.92
MDL Number MFCD01074545
InChI Key QLULGIRFKAWHOJ-UHFFFAOYSA-N
Synonym pyridine-4-boronic acid, 4-pyridineboronic acid, 4-pyridylboronic acid, pyridin-4-yl boronic acid, 4-pyridinylboronic acid, pyridin-4-yl-4-boronic acid, 4-pyridinebornic acid, pyridyl-4-boronic acid, 4-pyridinyl boronic acid, pyridine-4-ylboronic acid
PubChem CID 2734379
SMILES OB(O)C1=CC=NC=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
IUPAC Name pyridin-4-ylboronic acid
Formula Weight 122.92
Percent Purity 97%
Chemical Name or Material Pyridin-4-ylboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.