missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-4-carboximidamide hydrochloride, 95%, Thermo Scientific™

Product Code. 10748455
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10748455 50 g 50g
10712967 5 g 5g
10661483 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10748455 Supplier Thermo Scientific Maybridge Supplier No. CD05159EE
 Restricted Item
Estimated Shipment: 19-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 6345-27-3
Molecular Formula C6H8ClN3
Molecular Weight (g/mol) 157.60
MDL Number MFCD00040385
InChI Key IONKMFGAXKCLMI-UHFFFAOYSA-N
Synonym isonicotinimidamide hydrochloride, pyridine-4-carboximidamide hydrochloride, 4-amidinopyridine hydrochloride, isonicotinamidine hydrochloride, pyridine-4-carboxamidine hydrochloride, 4-pyridinecarboximidamide hydrochloride, 4-pyridinecarboximidamide, monohydrochloride, isonicotinamidine, hydrochloride, 4-amidino-pyridine hydrochloride, pyridine-4-carboxamidine, chloride
PubChem CID 2776844
IUPAC Name pyridine-4-carboximidamide;hydrochloride
SMILES Cl.NC(=N)C1=CC=NC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 50 g
Formula Weight 157.6
Percent Purity 95%
Chemical Name or Material Pyridine-4-carboximidamide hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.