missing translation for 'onlineSavingsMsg'
Learn More

Pyridine, 99+%, ACS reagent

Product Code. 10795253 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
Metal Drum
Unit Size:
1 L
2.5 L
25 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10795253 1 L Glass Bottle 1 L
10075970 500 mL EcoSafPak™ Glass Bottle 500mL
10243423 2.5 L Glass Bottle 2.5 L
10617954 25 L Metal Drum 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10795253 Supplier Thermo Scientific Acros Supplier No. 418540010
  • ONLINE EXCLUSIVE PRICE
    195.00 €
    189.00 € / 1 L
    Save 6.00 €
    3% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 110-86-1 | C5H5N | 79.102 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.102
MDL Number MFCD00011732
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
PubChem CID 1049
ChEBI CHEBI:16227
IUPAC Name pyridine
SMILES C1=CC=NC=C1

Specifications

Melting Point -42.0°C
Color Colorless
Density 0.9780g/mL
Quantity 1 L
Flash Point 17°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.5085 to 1.5105
Residue after Evaporation 0.002% max.
Assay Percent Range 99+%
Specific Gravity 0.978
Vapor Pressure Vapor Pressure: 20mbar at 20°C
Viscosity 0.95 mPa.s (20°C)
UN Number 1282
Water 0.1% max. (K.F.)
Beilstein 20, 181
Fieser 01,958; 02,349; 04,414; 06,497; 11,448; 12,416; 13,293; 15,1; 17,153
Merck Index 15, 8081
Boiling Point 115.0°C to 116.0°C
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and most other common, organic solvents
Formula Weight 79.1
Percent Purity ≥99%
Grade ACS
Chemical Name or Material Pyridine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Pyridine
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.