missing translation for 'onlineSavingsMsg'
Learn More

Pyridine, ACS Reagent, ≥99%, Honeywell™ Riedel-de Haën™

Product Code. 15605100
Click to view available options
Quantity:
100 mL
2.5 L
Packaging:
Glass Bottle
Unit Size:
100mL
2.5 L
This item is not returnable. View return policy

Product Code. 15605100

Brand: Honeywell-Riedel-de Haen 3605702.5L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ACS Reagent,≥99.0%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.102
MDL Number MFCD00011732
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
PubChem CID 1049
ChEBI CHEBI:16227
IUPAC Name pyridine
SMILES C1=CC=NC=C1

Specifications

pH 8.8 (at 20°C)
Melting Point -42°C
Color Colorless
Density 0.978g/mL at 25°C
Boiling Point 115°C
Quantity 2.5 L
Explosion Limit 1.8% to 12.4% (V)
Flash Point 17°C (62.6°F)
Ignition Point 899°F
Odor Unpleasant
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 10mmHg (13.2°C), 20mmHg (25°C)
UN Number UN1282
Beilstein 103233
Solubility Information Completely miscible in water
Formula Weight 79.1g/mol
Percent Purity ≥99%
Chemical Name or Material Pyridine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.