missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-d5, for NMR, contains 1 v/v% TMS, 99 atom % D

Product Code. 10562041 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
Packaging:
Glass Bottle
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10562041 5 mL Glass Bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10562041 Supplier Thermo Scientific Acros Supplier No. 214560050
  • ONLINE EXCLUSIVE PRICE
    98.40 €
    90.50 € / 5mL
    Save 7.90 €
    8% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 7291-22-7 | C5H5N | 84.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Color Colorless
Density 1.0500g/mL
Deuterium (D) 98.5% min.
Flash Point 20°C
Packaging Glass Bottle
Refractive Index 1.507
Water 0.1% max. (Coulometric)
Quantity 5 mL
CAS 7291-22-7, 75-76-3
Melting Point -41.0°C
Boiling Point 114.4°C
Molecular Formula C5H5N
MDL Number MFCD00044639
Beilstein 20, III, 2305
Synonym pyridine-d5, 2h5 pyridine, c5d5n, 2 h? pyridine, pyridine d5, pyridine,crude,light, pyridine, perdeutero, ∼2∼h_5_ pyridine, de85c
InChI Key JUJWROOIHBZHMG-RALIUCGRSA-N
SMILES [2H]C1=NC([2H])=C([2H])C([2H])=C1[2H]
IUPAC Name 2,3,4,5,6-pentadeuteriopyridine
Molecular Weight (g/mol) 84.13
PubChem CID 558519
Formula Weight 84.14
Grade NMR
Chemical Name or Material Pyridine-d5, 99 atom % D
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.