missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-d5, for NMR, packaged in 1.00mL ampoules, 99% atom D, ACROS Organics™

Product Code. 10687012 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 x 1.00 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10687012 10 x 1.00 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10687012 Supplier Thermo Scientific Acros Supplier No. 217360100
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Specifications

Physical Form Liquid
Color Colorless
Density 1.0500g/mL
Deuterium (D) 98.5% min.
Water 0.1% max. (Coulometric)
Quantity 10 x 1.00 mL
CAS 7291-22-7
Molecular Formula C5H5N
MDL Number MFCD00044639
Beilstein 20, III, 2305
Synonym pyridine-d5, 2h5 pyridine, c5d5n, 2 h? pyridine, pyridine d5, pyridine,crude,light, pyridine, perdeutero, ∼2∼h_5_ pyridine, de85c
InChI Key JUJWROOIHBZHMG-RALIUCGRSA-N
SMILES [2H]C1=NC([2H])=C([2H])C([2H])=C1[2H]
IUPAC Name (²H₅)pyridine
Molecular Weight (g/mol) 84.13
PubChem CID 558519
Formula Weight 84.14
Grade NMR
Chemical Name or Material Pyridine-d5, 99 atom % D
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.