missing translation for 'onlineSavingsMsg'
Learn More

Pyridine,Solstice

Product Code. 16891240
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
16891240 2.5 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16891240 Supplier Burdick & Jackson BioSyn Supplier No. BB3332.5L
  • ONLINE EXCLUSIVE PRICE
    794.00 €
    717.53 € / 1 L
    Save 76.47 €
    9% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

BioSyn™, for biosynthesis and applications requiring low water content, >99.8%

  • Ultra-low water specifications
  • Low amine specifications
  • Low particulate levels
  • High solvent purity
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-86-1
PubChem CID 1049
ChEBI CHEBI:16227

Specifications

Quantity 2.5 L
Flash Point 20 °C
Packaging Glass Bottle
Physical Form Liquid, Clear
Refractive Index 1.5085 - 1.5109
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.