missing translation for 'onlineSavingsMsg'
Learn More

Pyridine, Pure, Solution, 65% (v/v) In Water, Fisher Chemical

Product Code. 12686727 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass Bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12686727 500 mL Amber Glass Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 12686727 Supplier Fisher Chemical Supplier No. J/9014/08
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Specifications

pH 8.5
Melting Point -42°C
CAS 110-86-1
Color Yellow
Quantity 500 mL
Packaging Amber Glass Bottle
Physical Form Liquid
Molecular Formula C5H5N
Vapor Pressure 18mmHg at 20°C
MDL Number 11732
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
Boiling Point 115°C
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
SMILES C1=CC=NC=C1
IUPAC Name pyridine
Molecular Weight (g/mol) 79.102
PubChem CID 1049
ChEBI CHEBI:16227
Formula Weight 79.1g/mol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Pyridine solution, 65% v/v in water
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.